AMBH580645D5 Ambeed, Inc.

Isoquinoline-3-carboximidamide hydrochloride

  • MDL number MFCD12755569
  • Cas Number : 1179362-42-5
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-isoquinolinecarboximidamide hydrochloride
Inchi Code 1S/C10H9N3.ClH/c11-10(12)9-5-7-3-1-2-4-8(7)6-13-9;/h1-6H,(H3,11,12);1H
Inchi Key IMCAGIJSBQEATO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1179362-42-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1179362-42-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart