AMBH580645D9 Ambeed, Inc.

2,4-Dibromo-5-fluoropyridine

  • MDL number MFCD11114541
  • Cas Number : 1256805-06-7
  • Molecular Weight : 254.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1256805-06-7.html

Safety Information

MSDS https://www.ambeed.com/products/1256805-06-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart