AMBH580645E0 Ambeed, Inc.

Ethyl 4-amino-1H-imidazole-2-carboxylate

  • MDL number MFCD09263979
  • Cas Number : 83566-37-4
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/83566-37-4.html

Safety Information

MSDS https://www.ambeed.com/products/83566-37-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart