AMBH580645E3 Ambeed, Inc.

NVP-HDM 201

  • MDL number MFCD30502893
  • Cas Number : 1448867-41-1
  • Molecular Weight : 555.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/1448867-41-1.html

Safety Information

MSDS https://www.ambeed.com/products/1448867-41-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart