AMBH580645E8 Ambeed, Inc.

4-Methylphenyl 2,3,4,6-tetra-O-acetyl-1-thio-�D-glucopyranoside

  • MDL number N/A
  • Cas Number : 28244-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/28244-94-2.html

Safety Information

MSDS https://www.ambeed.com/products/28244-94-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart