AMBH580645ED Ambeed, Inc.

3-Methyl-1-p-tolyl-5-pyrazolone

  • MDL number MFCD00035708
  • Cas Number : 86-92-0
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129°C at N/A mmHg
Purity 97
Flash Point 141.8
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 310.9°C at 760 mmHg
Product Link https://www.ambeed.com/products/86-92-0.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/86-92-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart