AMBH580645F1 Ambeed, Inc.

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole

  • MDL number MFCD13181969
  • Cas Number : 1810038-58-4
  • Molecular Weight : 245.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1810038-58-4.html

Safety Information

MSDS https://www.ambeed.com/products/1810038-58-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart