AMBH580645F8 Ambeed, Inc.

4,5-Dimethoxyphthalonitrile

  • MDL number MFCD27931583
  • Cas Number : 88946-67-2
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/88946-67-2.html

Safety Information

MSDS https://www.ambeed.com/products/88946-67-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart