AMBH580645F9 Ambeed, Inc.

9H-Carbazole-1,8-diamine

  • MDL number MFCD18450117
  • Cas Number : 117110-85-7
  • Molecular Weight : 197.2358

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/117110-85-7.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352-P305+P351+P338-P332+P313-P337+P313-P362
MSDS https://www.ambeed.com/products/117110-85-7.html
pictograms GHS07
hstatements H315-H319-H320
Availability details for
Quantity
  • Availability
  • Add to Cart