AMBH580645FD Ambeed, Inc.

Benzo[f]quinoline

  • MDL number MFCD00022235
  • Cas Number : 85-02-9
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 90-93
Iupac Name benzo[f]quinoline
Inchi Code 1S/C13H9N/c1-2-5-11-10(4-1)7-8-13-12(11)6-3-9-14-13/h1-9H
Inchi Key HCAUQPZEWLULFJ-UHFFFAOYSA-N

Description

Melting Point 90-93
Purity 98%
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/85-02-9.html

Safety Information

signalword Danger
pcodes P280-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/85-02-9.html
pictograms GHS05,GHS06
hstatements H301-H318
Availability details for
Quantity
  • Availability
  • Add to Cart