AMBH58064601 Ambeed, Inc.

N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD09878608
  • Cas Number : 477600-74-1
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/477600-74-1.html

Safety Information

MSDS https://www.ambeed.com/products/477600-74-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart