AMBH58064606 Ambeed, Inc.

5-(tert-Butyl)-1H-benzo[d]imidazole

  • MDL number MFCD19373985
  • Cas Number : 110534-15-1
  • Molecular Weight : 174.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/110534-15-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/110534-15-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart