AMBH5806460A Ambeed, Inc.

6-Fluoro-1-methyl-1H-indazole-4-carboxylic acid

  • MDL number MFCD20645868
  • Cas Number : 697739-07-4
  • Molecular Weight : 194.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/697739-07-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/697739-07-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart