AMBH58064610 Ambeed, Inc.

Copper(II) phthalocyanine

  • MDL number MFCD00010719
  • Cas Number : 147-14-8
  • Molecular Weight : 576.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 600�t N/A mmHg
Iupac Name copper(II) phthalocyanine-29,31-diide
Inchi Code 1S/C32H16N8.Cu/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2
Inchi Key XCJYREBRNVKWGJ-UHFFFAOYSA-N

Description

Melting Point 600�t N/A mmHg
Purity 98%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/147-14-8.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/147-14-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart