AMBH58064613 Ambeed, Inc.

Methyl 5-chloropyrimidine-2-carboxylate

  • MDL number MFCD16611603
  • Cas Number : 894797-00-3
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/894797-00-3.html

Safety Information

MSDS https://www.ambeed.com/products/894797-00-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart