AMBH58064615 Ambeed, Inc.

1-Bromo-2-(difluoromethyl)-4-fluorobenzene

  • MDL number MFCD14698532
  • Cas Number : 1198171-18-4
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1198171-18-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1198171-18-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart