AMBH5806461B Ambeed, Inc.

(3-Chloro-4-methylphenyl)(methyl)sulfane

  • MDL number MFCD11617781
  • Cas Number : 53250-85-4
  • Molecular Weight : 172.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-1-methyl-4-(methylsulfanyl)benzene
Inchi Code 1S/C8H9ClS/c1-6-3-4-7(10-2)5-8(6)9/h3-5H,1-2H3
Inchi Key WHADXSMKJDNHKP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/53250-85-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/53250-85-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart