AMBH5806461D Ambeed, Inc.

(R)-tert-Butyl (3-hydroxy-1-(methoxy(methyl)amino)-1-oxopropan-2-yl)carbamate

  • MDL number MFCD11041265
  • Cas Number : 167102-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/167102-61-6.html

Safety Information

MSDS https://www.ambeed.com/products/167102-61-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart