AMBH5806461E Ambeed, Inc.

1-Benzylazocan-5-one

  • MDL number MFCD23701698
  • Cas Number : 16853-08-0
  • Molecular Weight : 217.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/16853-08-0.html

Safety Information

MSDS https://www.ambeed.com/products/16853-08-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart