AMBH5806461F Ambeed, Inc.

Dibenzyl diselenide

  • MDL number MFCD00004767
  • Cas Number : 1482-82-2
  • Molecular Weight : 340.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 91-93�t N/A mmHg
bp 427.3�t 760 mmHg
Iupac Name 1,2-dibenzyldiselane
Inchi Code 1S/C14H14Se2/c1-3-7-13(8-4-1)11-15-16-12-14-9-5-2-6-10-14/h1-10H,11-12H2
Inchi Key HYAVEDMFTNAZQE-UHFFFAOYSA-N

Description

Melting Point 91-93�t N/A mmHg
Purity 95%
Flash Point 212.2
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 427.3�t 760 mmHg
Product Link https://www.ambeed.com/products/1482-82-2.html

Safety Information

signalword Danger
pcodes P260-P264-P270-P271-P273-P301+P310+P330-P304+P340+P311-P314-P391-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1482-82-2.html
pictograms GHS06,GHS08,GHS09
hstatements H301+H331-H373-H410
Availability details for
Quantity
  • Availability
  • Add to Cart