AMBH58064623 Ambeed, Inc.

2,3,5-Trifluorophenol

  • MDL number MFCD00075416
  • Cas Number : 2268-15-7
  • Molecular Weight : 148.0826

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2268-15-7.html

Safety Information

signalword Danger
pcodes P210-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/2268-15-7.html
pictograms GHS02,GHS07
hstatements H226-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart