AMBH58064626 Ambeed, Inc.

Pyridine-4-aldoxime

  • MDL number MFCD00006446
  • Cas Number : 696-54-8
  • Molecular Weight : 122.1246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/696-54-8.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/696-54-8.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart