AMBH5806462E Ambeed, Inc.

Ethyl 3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-�D-thioglucopyranoside

  • MDL number N/A
  • Cas Number : 99409-32-2
  • Molecular Weight : 479.5002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/99409-32-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/99409-32-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart