AMBH5806462F Ambeed, Inc.

2-Amino-5-bromo-4-fluorobenzamide

  • MDL number MFCD14585387
  • Cas Number : 1206969-09-6
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1206969-09-6.html

Safety Information

MSDS https://www.ambeed.com/products/1206969-09-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart