AMBH58064634 Ambeed, Inc.

Ammonium molybdate tetrahydrate

  • MDL number MFCD00167059
  • Cas Number : 12054-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/12054-85-2.html

Safety Information

MSDS https://www.ambeed.com/products/12054-85-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart