AMBH5806463D Ambeed, Inc.

2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine

  • MDL number MFCD06858479
  • Cas Number : 496-13-9
  • Molecular Weight : 120.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/496-13-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/496-13-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart