AMBH5806463E Ambeed, Inc.

4-Methoxybenzenesulfonic acid

  • MDL number MFCD08685124
  • Cas Number : 5857-42-1
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 86-88
bp 193-194�t 1 mmHg
Iupac Name 4-methoxybenzenesulfonic acid
Inchi Code 1S/C7H8O4S/c1-11-6-2-4-7(5-3-6)12(8,9)10/h2-5H,1H3,(H,8,9,10)
Inchi Key IWYVYUZADLIDEY-UHFFFAOYSA-N

Description

Melting Point 86-88
Purity 98%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 193-194�t 1 mmHg
Product Link https://www.ambeed.com/products/5857-42-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/5857-42-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart