AMBH58064649 Ambeed, Inc.

[1,1'-Biphenyl]-2,2'-diyldiboronic acid

  • MDL number MFCD21603686
  • Cas Number : 312968-33-5
  • Molecular Weight : 241.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name biphenyl-2,2'-diyldiboronic acid
Inchi Code 1S/C12H12B2O4/c15-13(16)11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(17)18/h1-8,15-18H
Inchi Key BVFQKUXUNMZSLZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/312968-33-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/312968-33-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart