AMBH5806464A Ambeed, Inc.

6-Chloro-3-methyl-3H-imidazo[4,5-c]pyridine

  • MDL number MFCD26743555
  • Cas Number : 7205-45-0
  • Molecular Weight : 167.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7205-45-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7205-45-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart