AMBH5806464E Ambeed, Inc.

(S)-2-(2-(2,4-Dichlorophenoxy)acetamido)pentanedioic acid

  • MDL number MFCD01725217
  • Cas Number : 32773-59-4
  • Molecular Weight : 350.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/32773-59-4.html

Safety Information

MSDS https://www.ambeed.com/products/32773-59-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart