AMBH58064653 Ambeed, Inc.

Potassium trifluoro(pentyl)borate

  • MDL number MFCD07368249
  • Cas Number : 872054-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/872054-60-9.html

Safety Information

MSDS https://www.ambeed.com/products/872054-60-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart