AMBH58064659 Ambeed, Inc.

(R)-2-(Diphenyl((trimethylsilyl)oxy)methyl)pyrrolidine

  • MDL number MFCD09265100
  • Cas Number : 943757-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/943757-71-9.html

Safety Information

MSDS https://www.ambeed.com/products/943757-71-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart