AMBH5806465A Ambeed, Inc.

N-(o-Tolyl)acetamide

  • MDL number MFCD00014961
  • Cas Number : 120-66-1
  • Molecular Weight : 149.1897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/120-66-1.html

Safety Information

signalword Warning
pcodes P264-P270-P280-P301+P312+P330-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P501
MSDS https://www.ambeed.com/products/120-66-1.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart