AMBH5806466C Ambeed, Inc.

(1H-Pyrazolo[3,4-b]pyridin-5-yl)boronic acid

  • MDL number MFCD18250847
  • Cas Number : 1417985-25-1
  • Molecular Weight : 162.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1417985-25-1.html

Safety Information

MSDS https://www.ambeed.com/products/1417985-25-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart