AMBH58064672 Ambeed, Inc.

(2S,4S)-4-((tert-Butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08704525
  • Cas Number : 1279034-78-4
  • Molecular Weight : 230.2609

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1279034-78-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1279034-78-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart