AMBH58064673 Ambeed, Inc.

(E)-5-(4-hydroxy-3-methoxystyryl)benzene-1,3-diol

  • MDL number MFCD12407151
  • Cas Number : 32507-66-7
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8
Country of Origin China
Product Link https://www.ambeed.com/products/32507-66-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/32507-66-7.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart