AMBH58064675 Ambeed, Inc.

Ethyl 3-(methylamino)-3-oxopropanoate

  • MDL number MFCD08458724
  • Cas Number : 71510-95-7
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/71510-95-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/71510-95-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart