AMBH58064676 Ambeed, Inc.

1,3-Dimethyl-1H-pyrazol-4-ol

  • MDL number MFCD23110698
  • Cas Number : 85985-66-6
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/85985-66-6.html

Safety Information

MSDS https://www.ambeed.com/products/85985-66-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart