AMBH58064688 Ambeed, Inc.

Ammonium molybdate

  • MDL number MFCD00011596
  • Cas Number : 13106-76-8
  • Molecular Weight : 196.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.9
Country of Origin China
Product Link https://www.ambeed.com/products/13106-76-8.html

Safety Information

signalword Danger
pcodes P261-P273-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13106-76-8.html
pictograms GHS07,GHS08
hstatements H302-H315-H319-H335-H351-H412
Availability details for
Quantity
  • Availability
  • Add to Cart