AMBH5806468A Ambeed, Inc.

Methyl 4-amino-2-chloro-5-methoxybenzoate

  • MDL number MFCD23707454
  • Cas Number : 1427390-51-9
  • Molecular Weight : 215.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-amino-2-chloro-5-methoxybenzoate
Inchi Code 1S/C9H10ClNO3/c1-13-8-3-5(9(12)14-2)6(10)4-7(8)11/h3-4H,11H2,1-2H3
Inchi Key SXTDGPPXGMTYEP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1427390-51-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1427390-51-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart