AMBH58064693 Ambeed, Inc.

trans-3-Hexene

  • MDL number MFCD00009386
  • Cas Number : 13269-52-8
  • Molecular Weight : 84.1595

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13269-52-8.html

Safety Information

signalword Danger
pcodes P210-P301+P310-P331
MSDS https://www.ambeed.com/products/13269-52-8.html
pictograms GHS02,GHS08
hstatements H225-H304
Availability details for
Quantity
  • Availability
  • Add to Cart