AMBH580646A1 Ambeed, Inc.

(S)-tert-Butyl 3-methoxypyrrolidine-1-carboxylate

  • MDL number MFCD16620895
  • Cas Number : 550371-69-2
  • Molecular Weight : 201.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/550371-69-2.html

Safety Information

MSDS https://www.ambeed.com/products/550371-69-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart