AMBH580646B2 Ambeed, Inc.

1-Allylcyclopropane-1-sulfonyl chloride

  • MDL number MFCD11046295
  • Cas Number : 923032-59-1
  • Molecular Weight : 180.6500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/923032-59-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/923032-59-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart