AMBH580646B6 Ambeed, Inc.

YH239-EE

  • MDL number MFCD28099816
  • Cas Number : 1364488-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1364488-67-4.html

Safety Information

MSDS https://www.ambeed.com/products/1364488-67-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart