AMBH580646C5 Ambeed, Inc.

5-Aminotetrazole monohydrate

  • MDL number MFCD00149327
  • Cas Number : 15454-54-3
  • Molecular Weight : 103.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198°C
Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/15454-54-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15454-54-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart