AMBH580646C8 Ambeed, Inc.

Propargyl ALPHA-D-mannopyranoside

  • MDL number N/A
  • Cas Number : 854262-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/854262-01-4.html

Safety Information

MSDS https://www.ambeed.com/products/854262-01-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart