AMBH580646CE Ambeed, Inc.

2-Iodo-4-nitro-1-(trifluoromethoxy)benzene

  • MDL number MFCD13194454
  • Cas Number : 194344-29-1
  • Molecular Weight : 333.0033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/194344-29-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/194344-29-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart