AMBH580646D7 Ambeed, Inc.

5-Methylisoxazol-4-amine

  • MDL number MFCD11848435
  • Cas Number : 87988-94-1
  • Molecular Weight : 98.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/87988-94-1.html

Safety Information

MSDS https://www.ambeed.com/products/87988-94-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart