AMBH580646DF Ambeed, Inc.

3,3,4-Trimethylpiperazin-2-one

  • MDL number MFCD19216480
  • Cas Number : 86290-99-5
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/86290-99-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/86290-99-5.html
pictograms GHS07
hstatements H302-H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart