AMBH580646E5 Ambeed, Inc.

1,4-Diphenethylbenzene

  • MDL number MFCD12407076
  • Cas Number : 1985-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1985-58-6.html

Safety Information

MSDS https://www.ambeed.com/products/1985-58-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart